Learn More
Diba Omnifit™ Luer Adapter
Adapters fit into 1/4 in.–28 UNF flat bottom ports in most Omnifit connectors and valves
Brand: Diba 002502
Code : Z3
Additional Details : Weight : 0.50000kg
Description
- Provide quick, push-on connection for softwall tubing or luer
- Made from chemically resistant ETFE
- Autoclavable
- OF001206 features a thread to male Luer lock adapter with a polypropylene lock ring
- OF002501 features male luer fittings to 1/4 in.–28 UNF thread
- OF002502 features femal luer fittings to 1/4 in.–28 UNF thread
Specifications
Female Luer Fitting | |
ETFE |