Learn More
asecos™ Oil Sorbents Mat
The Oil Sorbents Mat is water repellant for the safe absorption of oils, petrol, diesel, etc.
Brand: asecos™ 11327
This item is not returnable.
View return policy
Description
- Very strong durability
- Suitable for heavy and long-lasting use
- Tightly woven fibres, high abrasion resistance and low-lint surface
- Outstanding absorption performance
- Large absorption capacity (up to 16 time of own weight)
- Light for smaller leakages and prevention
- Heavy for larger leakages or absorption of puddles
Specifications
Blue | |
Oil Only Sorbent Pad | |
400.00 mm | |
400.00 mm | |
7 kg |
Heavy-duty Cloth | |
15.75 in. | |
15.75 in. | |
15.43 lb. |