Learn More
asecos™ Oil Sorbents Mat
The Oil Sorbents Mat is water repellant for the safe absorption of oils, petrol, diesel, etc.
Brand:  asecos™ 9934
 View more versions of this product
Product Code. 17836516
  • £191.00 / Pack of 100
Estimated Shipment: 13-01-2025
to see stock.
Explore more special offers
Add to basket
Description
Description
  • Very strong durability
  • Suitable for heavy and long-lasting use
  • Tightly woven fibres, high abrasion resistance and low-lint surface
  • Outstanding absorption performance
  • Large absorption capacity (up to 16 time of own weight)
  • Light for smaller leakages and prevention
  • Heavy for larger leakages or absorption of puddles
Specifications
Specifications
Blue
Oil Only Sorbent Pad
400.00 mm
400.00 mm
8 kg
Heavy-duty Cloth
15.75 in.
15.75 in.
17.64 lb.