Learn More
Chemical Identifiers
CAS | 124-13-0 |
---|---|
Molecular Formula | C8H16O |
Molecular Weight (g/mol) | 128.22 |
MDL Number | MFCD00007029 |
InChI Key | NUJGJRNETVAIRJ-UHFFFAOYSA-N |
Synonym | caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal |
PubChem CID | 454 |
ChEBI | CHEBI:17935 |
IUPAC Name | octanal |
SMILES | CCCCCCCC=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10285220
|
Thermo Scientific Acros
129481000 |
100 mL | Glass bottle |
£20.00
100mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10725231
|
Thermo Scientific Acros
129485000 |
500 mL | Glass bottle |
£43.42
500mL |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
124-13-0 | |
128.22 | |
NUJGJRNETVAIRJ-UHFFFAOYSA-N | |
454 | |
octanal |
C8H16O | |
MFCD00007029 | |
caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal | |
CHEBI:17935 | |
CCCCCCCC=O |
Specifications
124-13-0 | |
100.0 | |
Colorless to Yellow | |
171.0°C | |
Authentic | |
C8H16O | |
1.4170 to 1.4200 | |
01, 704 | |
0.821 | |
Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and mineral oil, insoluble in glycerol | |
CCCCCCCC=O | |
128.22 | |
CHEBI:17935 | |
99% | |
Octyl aldehyde, 99% |
98.5 | |
12.0°C to 15.0°C | |
0.8210g/mL | |
51°C | |
98.5% min. (GC) | |
CH3(CH2)6CHO | |
MFCD00007029 | |
15, 1768 | |
caprylic aldehyde, caprylaldehyde, n-octanal, 1-octanal, n-octyl aldehyde, n-octaldehyde, n-caprylaldehyde, octanaldehyde, aldehyde c-8, n-octylal | |
NUJGJRNETVAIRJ-UHFFFAOYSA-N | |
octanal | |
454 | |
128.21 | |
Liquid |
Safety and Handling
GHS H Statement
Flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove conta
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 204-683-8
missing translation for 'rtecsNumber' : RG7780000
missing translation for 'tsca' : TSCA
- Octyl aldehyde
Signal Word
- Warning
Hazard Category
- LONG-TERM AQUATIC HAZARD Chronic 2
- Serious eye damage/eye irritation Category 2
- Flammable liquid Category 3
- Skin corrosion/irritation Category 2
Hazard Statement
- H315-Causes skin irritation.
- H319-Causes serious eye irritation.
- H411-Toxic to aquatic life with long lasting effects.
- H226-Flammable liquid and vapour.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P362-Take off contaminated clothing.
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.