missing translation for 'onlineSavingsMsg'
Learn More

o-Tolylacetyl chloride, 98%, Thermo Scientific™

£43.00 - £123.00

Chemical Identifiers

CAS 10166-09-3
Molecular Formula C9H9ClO
Molecular Weight (g/mol) 168.62
MDL Number MFCD03424714
InChI Key FKMIBYMTHOJWCY-UHFFFAOYSA-N
Synonym o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
PubChem CID 2760632
IUPAC Name 2-(2-methylphenyl)acetyl chloride
SMILES CC1=CC=CC=C1CC(=O)Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15497767
View Documents
Thermo Scientific Alfa Aesar
H37418.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
15487767
View Documents
Thermo Scientific Alfa Aesar
H37418.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

10166-09-3
168.62
FKMIBYMTHOJWCY-UHFFFAOYSA-N
2760632
CC1=CC=CC=C1CC(=O)Cl
C9H9ClO
MFCD03424714
o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
2-(2-methylphenyl)acetyl chloride

Specifications

10166-09-3
84°C (1.5 mmHg)
MFCD03424714
Moisture sensitive
FKMIBYMTHOJWCY-UHFFFAOYSA-N
2-(2-methylphenyl)acetyl chloride
2760632
98%
o-Tolylacetyl chloride
Pink
C9H9ClO
UN3265
o-tolyl-acetyl chloride, o-tolylacetyl chloride, 2-2-methylphenyl acetyl chloride, o-methylphenylacetyl chloride, 2-methylphenylacetyl chloride, 2-methylbenzeneacetyl chloride, 2-methylphenyl acetyl chloride, 2-methylphenylacetic acid chloride
CC1=CC=CC=C1CC(=O)Cl
168.62
168.62
Liquid
Videos
Seguridad y manipulación

Seguridad y manipulación

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packaging Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures; Store under Nitrogen

SDS
Documentos

Documentos

Certificados