missing translation for 'onlineSavingsMsg'
Learn More

o-Phthaloyl dichloride, 90%, remainder mainly phthalic acid, Thermo Scientific Chemicals

Product Code. 10031400
Click to view available options
Quantity:
5 mL
100 mL
500 mL
Packaging:
Glass Bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-95-9
MDL Number MFCD00000666
InChI Key FYXKZNLBZKRYSS-UHFFFAOYSA-N
Synonym phthaloyl chloride, phthaloyl dichloride, phthalic chloride, phthalyl chloride, phthalic dichloride, phthalic acid dichloride, phthalyl dichloride, 1,2-benzenedicarbonyl dichloride, o-phthaloyl dichloride, phthaloyldichloride
PubChem CID 6955
IUPAC Name benzene-1,2-dicarbonyl chloride
SMILES C1=CC=C(C(=C1)C(=O)Cl)C(=O)Cl

Specifications

Melting Point 12.0°C
Color Yellow
Density 1.4000g/mL
Boiling Point 269.0°C to 271.0°C
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 90%
Packaging Glass Bottle
Molecular Formula C8H4Cl2O2
Linear Formula C6H4(COCl)2
Refractive Index 1.5674 to 1.5694
Quantity 100 mL
Beilstein 09, 805
Fieser 01,882; 14,263
Merck Index 15, 7487
Specific Gravity 1.4
Solubility Information Solubility in water: decomposes. Other solubilities: soluble in ether, decomposes in alcohol
Molecular Weight (g/mol) 203.018
Formula Weight 203.02
Percent Purity 90%
Physical Form Liquid
Chemical Name or Material o-Phthaloyl dichloride, Remainder mainly phthalic acid
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'