missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NUP62CL Rabbit anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 25381-1-AP-150UL
This item is not returnable.
View return policy
Specifications
NUP62CL | |
Polyclonal | |
Unconjugated | |
NUP62CL | |
NUP62CL, NUP62L | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
54830 | |
-20°C | |
Liquid |
Western Blot | |
0.36 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9H1M0 | |
NUP62CL | |
NUP62CL Fusion Protein Ag21107 | |
150 μL | |
Primary | |
Human | |
Antibody | |
IgG |