missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ NucleoMag Plant kit for DNA purification from plants

Flexible magnetic bead based isolation of DNA from a multitude of plant samples.

£658.00 - £3485.00

Specifications

Capacity 0.4 μg/μL beads
Isolation Technology Magnetic Bead
For Use With (Application) For Manual and Automated Isolation of DNA
Product Type NucleoMag™ 96 C-Bead
Test Time 120min.
View More Specs

Products 2
Product Code Brand No. of Reactions Price Quantity & Availability  
Product Code Brand No. of Reactions Price Quantity & Availability  
12795224
View accessory products View Documents
Macherey-Nagel™
744400.24
24 x 96
£3485.00
2304 reactions
In Stock
to see stock.
Add to basket
 
12728672
View accessory products View Documents
Macherey-Nagel™
744400.4
4 x 96
£658.00
384 reactions
Please to purchase this item. Need a web account? Register with us today!
Description

Description

After lysis of the blood cells genomic DNA is selectively bound to the NucleoMag Blood beads. Contaminants are washed away and the DNA can be eluted from the beads under low-salt conditions.As the magnetic bead technology provides a closed system (no transfer of sample from one well to another is necessary during the preparation) the risk for cross-contamination is greatly reduced. The NucleoMag technology can be easily adapted to common laboratory automation platforms. It can be used either with static-pin separators (eg. NucleoMag SEP) or magnetic separators integrated into robotic workstations.Kit components: NucleoMag B-Beads, elution plate (u-bottom), buffers. For further information on this product please contact our Technical Support team.
  • Magnetic bead technology
  • Yield: 1μg to 4μg
  • Sample material: 100μL whole blood
  • Elution volume: 25μL to 100μL
  • Binding capacity: 0.4μL/1μL beads
  • Preparation time: 120 min./96 preps
  • Fragment size: 300bp to 30kb
  • Format: superparamagnetic beads (size of beads: 1μm to 3μm)
For manual or automated isolation of genomic DNA from whole blood.
Specifications

Specifications

0.4 μg/μL beads
For Manual and Automated Isolation of DNA
120min.
Highly Reactive Superparamagnetic Beads
Magnetic Bead
NucleoMag™ 96 C-Bead
10 to 20μg
Videos
Safety and Handling

Safety and Handling

Signal Word
  • Danger

Hazard Category
  • Flammable liquid Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Serious eye damage/eye irritation Category 2
  • Specific target organ toxicity Category 3
  • Respiratory sensitiser Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H226-Flammable liquid and vapour.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H336-May cause drowsiness or dizziness.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P233-Keep container tightly closed.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .

Supplemental information
  • MIXTURE LIST-Contains : dodecyl sulfate, sodium salt / 2-propanol / ethanol, sodiumperchlorate

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.