missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoBond™ Xtra Maxi Plus
Purify transfection-grade plasmid DNA with high yield. The NucleoBond™ Xtra Maxi Plus Kit represents the new generation of anion exchanger allowing high yield, ultra-fast. The Plus kit contains NucleoBond™ Finalizers to speed up DNA precipitation.
£86.50 - £460.00
Specifications
Buffer | Yes |
---|---|
Format | Maxi Gravity-flow Column |
Isolation Technology | Anion Exchange Chromatography Technology |
RNase | Yes |
Size | <300kbp |
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | No. of Reactions | Price | Quantity & Availability | |||||
12758412
|
Macherey-Nagel™
740416.10 |
10 |
£460.00
10 reactions |
Estimated Shipment: 23-07-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11915452
|
Macherey-Nagel™
740416.2 |
2 |
£86.50
2 reactions |
Estimated Shipment: 02-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Features
- Proven anion-exchange technology – transfection-grade plasmid DNA
- Patented endotoxin removal by additional washing step – no extra ice incubation necessary
- Optimized column design – Maxi prep in 35 min.
- Column filter included – high filter flow-rates, parallel lysate clearing and loading onto the column
- Improved silica material – typical yields of 1000μg (Maxi)
- NucleoBond™ Xtra EF kits contain NucleoBond™ Finalizers to speed up DNA precipitation
Applications
- High-copy plasmids from E. coli
- Low-copy plasmids from E. coli (increased buffer volumes required)
Specifications
Yes | |
Anion Exchange Chromatography Technology | |
<300kbp |
Maxi Gravity-flow Column | |
Yes | |
1000μg |