missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NTRK1 (TRKA), His Tag Recombinant Protein
Human NTRK1 (TRKA), His Tag Recombinant Protein
Brand: Invitrogen PV4114
Code : 50
Additional Details : Weight : 0.05250kg
Description
NTRK1 (TRKA) belongs to the neurotrophic factor family of related polypeptides central to the development and maintenance of the mammalian nervous system.Specifications
NP_002520 | |
42.8 kDa | |
NTRK1 (TRKA), TRKA, TRK, MTK | |
Human | |
His Tag | |
Tyrosine Kinases | |
Baculovirus | |
Recombinant, Cytoplasmic | |
Signal Transduction |
Kinase Assay | |
1mg | |
Neurotrophic Receptor (NTRK) Kinase Family | |
Recombinant | |
Storage Buffer: 50mM Tris, pH 7.5, 100mM NaCl, 0.2mM EDTA, 0.05% Triton X-100, 5mM DTT, 50% glycerol Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. |
|
Dry Ice | |
Kinases & Inhibitors | |
Liquid |
For Research Use Only. Not for use in diagnostic procedures.