Learn More
NPRL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA558289
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: KLIQFGLMKN LIRRLQKYPV RVTREEQSHP ARLYTGCHSY DEICCKTGMS YHELDERLEN DPNIIICW Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
NPRL2 Polyclonal antibody specifically detects NPRL2 in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
NPRL2 | |
Polyclonal | |
Unconjugated | |
NPRL2 | |
2810446G01Rik; G21; G21 protein; GATOR complex protein NPRL2; Gene 21 protein; homologous to yeast nitrogen permease (candidate tumor suppressor); nitrogen permase homolog; nitrogen permease regulator 2-like protein; nitrogen permease regulator-like 2; nitrogen permease regulator-like 2 (S. cerevisiae); NPR 2L; NPR L2; NPR like 2; NPR2; NPR2 like; NPR2L; NPR2-like protein; NPR2-like, GATOR1 complex subunit; NPRL 2; Nprl2; Tumor suppressor candidate 4; TUSC 4; TUSC 4 protein; TUSC4; TUSC4 protein | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
10641 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q8WTW4 | |
NPRL2 | |
Recombinant protein corresponding to Human NPRL2 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only