missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NOXA2/p67phox Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15551-1-AP-20UL
This item is not returnable.
View return policy
Description
NCF2, NCF1, and a membrane bound cytochrome b558 are required for activation of the latent NADPH oxidase (necessary for superoxide production). Defects in NCF2 are a cause of chronic granulomatous disease autosomal recessive cytochrome-b-positive type 2 (CGD2). Chronic granulomatous disease is a genetically heterogeneous disorder characterized by the inability of neutrophils and phagocytes to kill microbes that they have ingested. Patients suffer from life-threatening bacterial/fungal infections.Specifications
NOXA2/p67phox | |
Polyclonal | |
Unconjugated | |
NCF2 | |
NADPH oxidase activator 2, NCF 2, NCF2, Neutrophil cytosol factor 2, neutrophil cytosolic factor 2, NOXA2, P67 PHOX, p67phox | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17970, 364018, 4688 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.29 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
A7E3N2, O70145, P19878 | |
Ncf2 | |
NOXA2/p67phox Fusion Protein Ag7909 | |
20 μL | |
Primary | |
Rat, Mouse, Human | |
Antibody | |
IgG |