missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ ZOOM™ Focusing Buffer pH 3-7
For use with ZOOM IPG Strips
Brand: Invitrogen™ ZB10037
This item is not returnable.
View return policy
Description
Included in Basic Protein Kit.
IsoElectric Focusing, Protein Sample Fractionation, Protein Sample Preparation and Protein Purification, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
The ZOOM™ IEF Fractionator Combo Kit includes the ZOOM™ IEF Fractionator; one pack ZOOM™ Disks of each pH; and ZOOM™ Reagents (urea, thiourea, CHAPS, carrier ampholytes pH 3-10, anode buffers, and cathode buffers). ZOOM™ Disks, Carrier Ampholytes, and Buffers are stored at +4°C. | |
IEF | |
Focusing Buffer | |
ZOOM™ | |
Wet Ice |
Focusing Buffers | |
3 to 7 | |
ZOOM™ IPG Strips | |
10 mL |
For Research Use Only. Not for use in diagnostic procedures.