Learn More
Novex™ Bolt™ Minigel Tank
Accommodates up to two gels per run
Brand: Invitrogen B4477599
Includes: Gel Runner Tank; Cassette Clamp (Left); Cassette Clamp (Right); Tank Base; Tank Lid; Gel Knife; Power Supply Adapters
Description
The Bolt Minigel Tank is compatible with Bolt Bis-Tris Plus gels. The unique tank design enables convenient side-by-side gel loading and enhanced viewing during use. Optimized conditions using constant voltage allow for Bolt Bis-Tris Plus gels to be run in approximately 35 minutes. Run times may vary depending on buffer conditions and power supplies being used. Bolt Minigel Tank is required for use of all Bolt gels.
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Specifications
Bolt | |
Gel Runner Tank; Cassette Clamp (Left); Cassette Clamp (Right); Tank Base; Tank Lid; Gel Knife; Power Supply Adapters |