missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BenchMark™ His-tagged Protein Standard

Used as a positive control and for molecular weight sizing in his-tagged fusion protein detection

Brand:  Invitrogen™ LC5606

Code : 50

Additional Details : Weight : 0.00454kg

Product Code. 10043462

  • £251.00 / Each
In Stock
to see stock.
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Includes: 125μL (25 applications of 5μL each) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, DTT, and Coomassie Blue G-250

Description

Description

The BenchMark His-tagged Protein Standard can be used with NuPAGE™ Gels and Tris-Glycine Gels.

  • Verification of his-tagged fusion protein detection using InVision™ In-Gel Stain or an Anti-His(C-term) antibody
  • Ten sharp and clear bands in the range of 10-160 kDa for molecular weight estimation of his-tagged proteins
  • Compatibility with Coomassie™ R-250 stain
  • A ready-to-use, time-saving format
  • Gel Compatibility: NovexTricine Gels, NovexTris-Glycine Gels, NuPAGE Bis-Tris Gels, SDS-PAGE Gels

1D Gel Electrophoresis, Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

Order Info

Shipping Condition: Dry Ice

Specifications

Specifications

125 μL (25 applications of 5 μL each) is provided in loading buffer consisting of Tris-HCl, SDS, Glycerol, DTT, and Coomassie™ Blue G-250.

Store at -20°C. Avoid repeated freezing and thawing.
10
Yes
Unstained
160, 120, 80, 60, 50, 40, 30, 20, 15, 10 kDa
125 μL
Western Blotting, SDS-PAGE
InVision™ His-tag In-gel Stain
Protein Ladder
10 to 160 kDa
Novex™ Tricine Gels, Novex™ Tris-Glycine Gels, NuPAGE™ Bis-Tris Gels, SDS-PAGE Gels
BenchMark™
Dry Ice
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.