missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex™ Zymogram, 10%, Tris-Glycine, 1.0 mm, Protein Gel, 10-well
Separation Range: 10 to 220 kd; Separation Type: Denaturing; For Use With: Mini Gel Tank, XCell SureLock Mini
Brand: Invitrogen™ EC6175BOX
Includes: Catalog numbers for gels refer to a single box. One box contains 10 gels.
Description
- Proteases are run on a gel containing one of these substrates under denaturing conditions and visualized as clear bands against a dark background following a simple renaturing, developing, and staining protocol
- Zymogram gels are commonly used to detect matrix metalloproteinases
- Tris-Glycine gels with a substrate (gelatin or casein) incorporated into the gel
- Made with high-purity, strictly quality-controlled reagents: Tris base, HCl, acrylamide, bisacrylamide, TEMED, APS, highly purified water and casein or gelatin as the substrate; All have a 4% stacking gel
Sample Loading Volume: 25μL; 1D Well
- The 10% Zymogram (Gelatin) Gel is a 10% Tris-Glycine gel with 0.1% gelatin as the substrate
- Sensitivity Level: 10% Zymogram Gelatin Gel: 5 x 10-6 units of collagenase
1D Gel Electrophoresis, Protein Gel Electrophoresis, Proteins, Expression, Isolation and Analysis
Order Info
Shipping Condition: Wet Ice
Specifications
XCell SureLock Mini-Cell | |
1.0 mm | |
Homogeneous Page | |
10 to 220 kDa | |
10-well | |
1D Well |
10% | |
Tris-Glycine | |
Protein | |
Denaturing | |
Each |
For Research Use Only. Not for use in diagnostic procedures.