Learn More
NK1.1 (CD161) Mouse anti-Mouse, PE, Clone: PK136, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech PE-65138-100UG
This item is not returnable.
View return policy
Description
The NK1.1 (CD161) antibody from Proteintech is a mouse monoclonal antibody generated with nk-1+ cells from mouse spleen and bone marrow of mouse NK1.1 (CD161). This antibody recognizes mouse antigen. The NK1.1 (CD161) antibody has been validated for the following applications: FC analysis. This antibody is pe.
Specifications
NK1.1 (CD161) | |
Monoclonal | |
0.2 mg/mL | |
Mouse | |
Primary | |
Mouse |
Flow Cytometry | |
PK136 | |
PE | |
100 μg | |
17057 | |
IgG2a |