missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nitrobenzene, 99%, Reagent Grade, Honeywell™
Click to view available options
Quantity:
1 L
2.5 L
10 L
Packaging:
Glass bottle
Stainless steel can
Description
- ≥95% for general laboratory use.
Specifications
Specifications
Melting Point | 5°C to 6°C |
Color | Yellow |
Density | 1.196g/mL at 25°C |
pH | 8.0 to 8.5 (at 1.0000g/L 20.0°C) |
Boiling Point | 210°C to 211°C |
Explosion Limit | 0.4 |
Flash Point | 88°C (190.4°F) |
Ignition Point | 899°F |
Odor | Pungent |
Packaging | Stainless steel can |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'