Learn More
Nickel pieces, 5x5cm (2.0x2.0in), 99.99% (metals basis), Thermo Scientific Chemicals
£110.55 - £355.77
Chemical Identifiers
CAS | 7440-02-0 |
---|---|
Molecular Formula | Ni |
Molecular Weight (g/mol) | 58.69 |
MDL Number | MFCD00011137 MFCD06798735 |
InChI Key | PXHVJJICTQNCMI-UHFFFAOYSA-N |
Synonym | raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum |
PubChem CID | 935 |
ChEBI | CHEBI:28112 |
IUPAC Name | nickel |
SMILES | [Ni] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11303647
|
Thermo Scientific Alfa Aesar
035482.36 |
500 g |
£110.55
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11313647
|
Thermo Scientific Alfa Aesar
035482.A3 |
2 kg |
£355.77
2kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7440-02-0 | |
58.69 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
935 | |
nickel |
Ni | |
MFCD00011137 MFCD06798735 | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
CHEBI:28112 | |
[Ni] |
Specifications
7440-02-0 | |
MFCD00011137 MFCD06798735 | |
PXHVJJICTQNCMI-UHFFFAOYSA-N | |
nickel | |
935 | |
99.99% | |
Pieces | |
Nickel pieces, 5x5cm (2.0x2.0 in.), Thickness ≈0.5 to 1.5cm |
Ni | |
raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum | |
[Ni] | |
58.69 | |
CHEBI:28112 | |
Odorless | |
(metals basis) |
Safety and Handling
GHS H Statement
H372-H351-H317
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
May cause an allergic skin reaction.
P201-P202-P260-P261-P264b-P270-P272-P280-P302+P352-P308+P313-P333+P313-P363-P501c
H317-H351-H372
missing translation for 'einecsNumber' : 231-111-4
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Nickel pieces
Signal Word
- Danger
Hazard Category
- Carcinogenicity Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H351-Suspected of causing cancer.
- H372-Causes damage to organs.
Precautionary Statement
- P201-Obtain special instructions before use.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/soap.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P362+P364-Take off contaminated clothing and wash it before reuse.
Supplemental information
- MIXTURE LIST-Contains : Nickel
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.