missing translation for 'onlineSavingsMsg'
Learn More

Nickel lump, vacuum remelted, low Oxygen, 99.97% (metals basis), Thermo Scientific Chemicals

Product Code. 11382038
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7440-02-0
Molecular Formula Ni
Molecular Weight (g/mol) 58.69
MDL Number MFCD00011137 MFCD06798735
InChI Key PXHVJJICTQNCMI-UHFFFAOYSA-N
Synonym raney alloy, fibrex, catalyst, particles, fibrex p, nickel, elemental, nichel italian, nickel, soluble salts, carbonyl powder, niccolum
PubChem CID 935
ChEBI CHEBI:28112
IUPAC Name nickel
SMILES [Ni]

Specifications

Quantity 100 g
Percent Purity 99.97%
Odor Odorless
Assay (metals basis)
Chemical Name or Material Nickel lump, vacuum remelted, Low Oxygen, Oxygen and Nitrogen analysis included

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'