missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) sulfate hexahydrate, Puriss. p.a., ACS Reagent, 99-102.0%, Honeywell Fluka™
Click to view available options
Quantity:
100 g
250 g
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Quantity | 250 g |
Linear Formula | NiSO4 · 6H2O |
UN Number | 3077 |
Formula Weight | 262.85g/mol |
Percent Purity | 99 to 102% |
Grade | ACS/puriss. p.a. |
Density | Vapor Density: 2.07 (vs air) |
Chemical Name or Material | Nickel(Ii) Sulfate Hexahydrate |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'