missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Nickel(II) Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™
Chemical Identifiers
CAS | 13478-00-7 |
---|---|
Molecular Formula | H12N2NiO12 |
Molecular Weight (g/mol) | 290.79 |
MDL Number | MFCD00149805 |
InChI Key | AOPCKOPZYFFEDA-UHFFFAOYSA-N |
Synonym | nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 |
PubChem CID | 61630 |
SMILES | O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Description
Nickel(II) Nitrate Hexahydrate, Extra Pure, SLR
Chemical Identifiers
13478-00-7 | |
290.79 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
61630 |
H12N2NiO12 | |
MFCD00149805 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O |
Specifications
137°C | |
13478-00-7 | |
5 | |
H12N2NiO12 | |
nickel nitrate hexahydrate, nickel ii nitrate hexahydrate, nickel dinitrate hexahydrate, nickel 2+ dinitrate hexahydrate, unii-xbt61wlt1j, nickelous nitrate hexahydrate, nickel 2+ nitrate, hexahydrate, xbt61wlt1j, nitric acid, nickel 2+ salt, hexahydrate, nickel ii nitrate, hexahydrate 1:2:6 | |
O.O.O.O.O.O.[Ni++].[O-][N+]([O-])=O.[O-][N+]([O-])=O | |
290.79 | |
290.8g/mol |
56.7°C | |
Blue-Green | |
Solid | |
MFCD00149805 | |
AOPCKOPZYFFEDA-UHFFFAOYSA-N | |
nickel(2+) hexahydrate dinitrate | |
61630 | |
Plastic powder jar |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Nickel(II) nitrate hexahydrate
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- LONG-TERM AQUATIC HAZARD Chronic 1
- ACUTE AQUATIC Acute 1
- Carcinogenicity Category 1A
- Serious eye damage/eye irritation Category 1
- Germ cell mutagenicity Category 2
- Oxidising solid Category 2
- Reproductive toxicity Category 1B
- Respiratory sensitiser Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
- Specific target organ toxicity Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
- H318-Causes serious eye damage.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H372-Causes damage to organs.
- H400-Very toxic to aquatic life.
- H410-Very toxic to aquatic life with long lasting effects.
- H350i-May cause cancer by inhalation.
- H360D-May damage the unborn child.
- H315-Causes skin irritation.
- H341-Suspected of causing genetic defects.
- H302-Harmful if swallowed.
- H332-Harmful if inhaled.
- H272-May intensify fire; oxidiser.
Precautionary Statement
- P201-Obtain special instructions before use.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P273-Avoid release to the environment.
- P281-Use personal protective equipment as required.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P405-Store locked up.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Nickel(II) Nitrate Hexahydrate, Extra Pure, SLR, Fisher Chemical™