missing translation for 'onlineSavingsMsg'
Learn More

Nickel(II) Chloride Hexahydrate, puriss. p.a., ≥98%, Honeywell Fluka™

Puriss. p.a.,≥98%

£194.67 - £351.01

Chemical Identifiers

CAS 7791-20-0
MDL Number MFCD00149809
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
15623460
View Documents
Honeywell-Fluka
31462-1KG
1 kg Poly Bottle
£194.67
1kg
Please to purchase this item. Need a web account? Register with us today!
15633460
View Documents
Honeywell-Fluka
31462-2.5KG
2.5 kg Poly Bottle
£351.01
2.50kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications

Chemical Identifiers

7791-20-0
MFCD00149809

Specifications

80°C
NiCl2 · 6H2O
UN3288
≥98%
3.550g/cm3
7791-20-0
MFCD00149809
237.69g/mol
puriss. p.a.
Nickel(Ii) Chloride Hexahydrate
Videos
Safety and Handling

Safety and Handling

Missing Phrase - N15.00950417-P201-P260-P280-P304 + P340 + P312-P308 + P313

H301 + H331-H315-H317-H334-H341-H350i-H360D-H372-H410

missing translation for 'dotInformation' : Class 6.1 - PG 3 - EHS - RQ - Toxic solid, inorganic, n.o.s., HI: all

missing translation for 'rtecsNumber' : QR6480000

SDS
Documents

Documents

Product Certifications