Learn More
Neutratek™ Neutral Detergent
For general manual cleaning, where acid or alkaline products cannot be used
Brand: Teknon TK140/5
Code : 04
Additional Details : Weight : 5.19000kg
Description
For general manual cleaning, where acid or alkaline products cannot be used. Neutratek is free of enzymes, sodium hypochlorite and phosphates, which can interfere with analytical methods. At between 0.2% to 2% concentration will effectively decontaminate non-ferrous metals, silica glass, delicate polymers and other delicate surfaces. To increase cleaning efficiency the solution can be heated and used in an ultrasonic bath.- High activity
- pH neutral
- Ultrasonic compatible
- Biodegradable A concentrated, neutral detergent for cleaning sensitive surfaces safely.
Specifications
Neutratek Neutral Detergent | |
For general manual cleaning, where acid or alkaline products cannot be used. | |
1 Ea. | |
5 L |
Safety and Handling
- Neutratek
Signal Word
- Danger
Hazard Category
- Serious eye damage/eye irritation Category 1
- Skin corrosion/irritation Category 2
- Skin sensitiser Category 1
Hazard Statement
- H318-Causes serious eye damage.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P405-Store locked up.