missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NEK7 Recombinant Human Protein

NEK7 belongs to the NIMA family of kinases.

Brand:  Invitrogen™ PR7441A

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15418593

  • £1604.00 / 100µg
Estimated Shipment: 11-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

NEK7 is involved in DNA replication and genotoxic stress response.

Specifications

Specifications

NP_598001.1
50mM Tris, pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% glycerol
100μg
RUO - research use only
NEK7
Human
GST Tag
Frozen
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology
63.7kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
NEK Kinase Family
Recombinant
Baculovirus
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers