missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ NEK2 Recombinant Human Protein
NEK2 is involved in cell cycle regulation.
Brand: Invitrogen™ PR4616A
Code : NEW
Additional Details : Weight : 0.01000kg
Description
NEK2 is essential for faithful chromosome segregation.
Specifications
NP_002488 | |
50mM Tris, pH 7.5, 500mM NaCl, 0.5mM EDTA, 0.05% Triton X-100, 2mM DTT, 50% glycerol | |
100μg | |
RUO - research use only | |
NEK2, NLK1 | |
Human | |
His Tag | |
Frozen |
Enzyme and Protein Activity Assay, Kinase Assay, Protein Assays and Analysis, Protein Biology | |
54.5kDa | |
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws. | |
Full Length | |
NEK Kinase Family | |
Recombinant | |
Baculovirus |