Learn More
NDUFS1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 12444-1-AP-20UL
 View more versions of this product
Product Code. 16810164
  • £135.00 / 20µL
Explore more special offers
Description
Description
The protein encoded by this gene belongs to the complex I 75 kDa subunit family. Mammalian complex I is composed of 45 different subunits. It locates at the mitochondrial inner membrane. This protein has NADH dehydrogenase activity and oxidoreductase activity. It transfers electrons from NADH to the respiratory chain. The immediate electron acceptor for the enzyme is believed to be ubiquinone. This protein is the largest subunit of complex I and it is a component of the iron-sulfur fragment of the enzyme. It may form part of the active site crevice where NADH is oxidized. Mutations in this gene are associated with complex I deficiency. Several transcript variants encoding different isoforms have been found for this gene.
Specifications
Specifications
NDUFS1
Polyclonal
Unconjugated
NDUFS1
CI 75Kd, Complex I 75kD, NDUFS1, PRO1304
Rabbit
Antigen Affinity Chromatography
RUO
227197, 301458, 4719
-20°C
Liquid
Western Blot, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Immunocytochemistry
0.17 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P28331, Q66HF1, Q91VD9
NDUFS1
NDUFS1 Fusion Protein Ag3135
20 μL
Primary
Rat, Human, Mouse