missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCKIPSD Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 11367-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is localized exclusively in the cell nucleus. It plays a role in signal transduction, and may function in the maintece of sarcomeres and in the assembly of myofibrils into sarcomeres. It also plays an important role in stress fiber formation. The gene is involved in therapy-related leukemia by a chromosomal translocation t(3;11)(p21;q23) that involves this gene and the myeloid/lymphoid leukemia gene. Alternative splicing occurs in this locus and two transcript variants encoding distinct isoforms have been identified.Specifications
NCKIPSD | |
Polyclonal | |
Unconjugated | |
NCKIPSD | |
AF3P21, Dia interacting protein 1, DIP, DIP 1, DIP1, NCKIPSD, ORF1, SH3 adapter protein SPIN90, SPIN90, VIP54, WASLBP, WISH | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
51517, 80987 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.16 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q9ESJ4, Q9NZQ3 | |
NCKIPSD | |
NCKIPSD Fusion Protein Ag1915 | |
20 μL | |
Primary | |
Mouse, Human | |
Antibody | |
IgG |