missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NCAM1/CD56 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 14255-1-AP-150UL
This item is not returnable.
View return policy
Description
NCAM, as a member of the immunoglobulin superfamily of adhesion molecules is characterized by several immunoglobulin (Ig)-like domains. The extracellular part of NCAM consists of five of these Ig domains and two fibronectin type III homology regions. NCAM is encoded by a single copy gene composed of 26 exons. However, at least 20-30 distinct isoforms can be generated by alternative splicing and by posttranslational modifications, such as sialylation. During sialylation, polysialic acid (PSA) carbohydrates are attached to the extracellular part of NCAM. Through its extracellular region, NCAM mediates homophilic interactions. In addition, NCAM can also undergo heterophilic interactions by binding extracellular matrix components, such as laminin, or other cell adhesion molecules, such as integrins.Specifications
NCAM1/CD56 | |
Polyclonal | |
Unconjugated | |
NCAM1 | |
CD56, MSK39, N CAM 1, NCAM, NCAM 1, NCAM1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
17967, 24586, 4684 | |
-20°C | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P13591, P13595, P13596 | |
Ncam1 | |
NCAM1/CD56 Fusion Protein Ag5528 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |