Learn More
NBR1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA564993
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: HEGHHVVDEAP PPVVGAKRLA ARAGKKPLAH YSSLVRVLGS DMKTPEDPAV QSFPLVPCDT DQPQDKPPDW FTSYLETFR Highest antigen sequence identity to the following orthologs - mouse 75%, rat 73%.
The protein encoded by this gene was originally identified as an ovarian tumor antigen monitored in ovarian cancer. The encoded protein contains a B-box/coiled coil motif, which is present in many genes with transformation potential, but the function of this protein is unknown. This gene is located on a region of chromosome 17q21.1 that is in close proximity to tumor suppressor gene BRCA1. Three alternatively spliced variants encoding the same protein have been identified for this gene.Specifications
NBR1 | |
Polyclonal | |
Unconjugated | |
NBR1 | |
1A13B; 1A1-3B; B-box protein; Ca125; Cell migration-inducing gene 19 protein; IAI3B; KIAA0049; M17S2; Membrane component chromosome 17 surface marker 2; Membrane component chromosome 17 surface marker 2 homolog; membrane component, chromosome 17, surface marker 2 (ovarian carcinoma antigen CA125); MIG19; migration-inducing protein 19; mKIAA0049; Nbr1; NBR1, autophagy cargo receptor; neighbor of Brca1 gene 1; neighbor of BRCA1 gene 1 protein; Next to BRCA1 gene 1 protein; next to the Brca1; ovarian carcinoma antigen CA125; Protein 1A1-3B; RGD1311421 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
4077 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q14596 | |
NBR1 | |
Recombinant Human NBR1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only