missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ National Target LoVial™ Crimp-Top Vials—Wide-Opening 2mL, 12 x 32mm

Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.

Brand:  Thermo Scientific™ C4011-K1W

 View more versions of this product

Product Code. 12490058

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »

Explore more special offers
This item is not returnable. View return policy

Description

Description

Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.

  • Fit most autosamplers that accommodate the 12 × 32mm vial format
  • Borosilicate clear glass Type 1, Class A or 51A amber glass
  • Uniformly flat bottom for security with inserts
  • I-D™ vials feature write-on patch with graduations
  • Target Colorband™ crimp top vials feature color-coded graduations and patch
  • Vials with an 11mm crimp finish require aluminum seal closures
  • Crimp/snap vials can be used with either 11mm aluminum seals or Snap-It™ Snap Caps
TRUSTED_SUSTAINABILITY
Specifications

Specifications

2mL Clear Glass I-D Vial, Kimshield™ Deactivated
2 mL
1.8 mL
Kimshield Deactivated Clear Glass I-D Vial
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers