missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ National Target LoVial™ Crimp-Top Vials—Wide-Opening 2mL, 12 x 32mm
Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.
Brand: Thermo Scientific™ C4011-S9
This item is not returnable.
View return policy
Description
Thermo Scientific™ National Target LoVial Crimp-Top Vials are designed with a wide-opening for easier filling.
- Fit most autosamplers that accommodate the 12 × 32mm vial format
- Borosilicate clear glass Type 1, Class A or 51A amber glass
- Uniformly flat bottom for security with inserts
- I-D™ vials feature write-on patch with graduations
- Target Colorband™ crimp top vials feature color-coded graduations and patch
- Vials with an 11mm crimp finish require aluminum seal closures
- Crimp/snap vials can be used with either 11mm aluminum seals or Snap-It™ Snap Caps
Specifications
1.5mL High Recovery MicroVials, NSC Silanized | |
2 mL | |
1.5 mL |