missing translation for 'onlineSavingsMsg'
Learn More

Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester, 97%, Thermo Scientific Chemicals

Product Code. 15446986
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Nalpha, Nepsilon-Di-Boc-L-lysine N-succinimidyl ester is used as a local anesthetic.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30189-36-7
Molecular Formula C20H33N3O8
Molecular Weight (g/mol) 443.50
MDL Number MFCD00057898
InChI Key IQVLXQGNLCPZCL-UGPWUYPHNA-N
Synonym boc-lys boc-osu, s-2,5-dioxopyrrolidin-1-yl 2,6-bis tert-butoxycarbonyl amino hexanoate, n,n'-di-boc-l-lysine hydroxysuccinimide ester, n, a,n, a-di-boc-l-lysine hydroxysuccinimide ester, n,n-di-boc-l-lysine hydroxysuccinimide ester, boc-lys-boc-osu, pubchem14936, boc-lys boc-osu calc. based on dry substance, c/n, 2,5-dioxoazolidinyl 2s-2,6-bis tert-butoxy carbonylamino hexanoate, 2,5-dioxopyrrolidin-1-yl n∼2∼,n∼6∼-bis tert-butoxycarbonyl-l-lysinate
PubChem CID 13875534
IUPAC Name (2,5-dioxopyrrolidin-1-yl) (2S)-2,6-bis[(2-methylpropan-2-yl)oxycarbonylamino]hexanoate
SMILES CC(C)(C)OC(=O)NCCCC[C@H](NC(=O)OC(C)(C)C)C(=O)ON1C(=O)CCC1=O

Specifications

Melting Point 184°C
Color White
Beilstein 1559007
Solubility Information Slightly soluble in water.
Quantity 25 g
Formula Weight 443.49
Sensitivity Moisture sensitive
Percent Purity 97%
Physical Form Powder
Chemical Name or Material Nalpha,Nepsilon-Di-Boc-L-lysine N-succinimidyl ester

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'