Learn More
Thermo Scientific™ Nalgene™ Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure

Protect against breakage and contamination with museum-quality PMP jars that lose only 2% of formaldehyde per year.
£15.55 - £154.00
Specifications
Color | Clear |
---|---|
Includes | Screw Closure |
Material | PMP |
No. per Pack | 4 |
Clarity | Clear |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | |||||
10537511
![]() |
Thermo Scientific™
2117-0060 |
60 mL | 53 mm |
£15.55
Pack of 4 |
In Stock
Log in to see stock. ![]() |
|||||
10547511
![]() |
Thermo Scientific™
2117-0125 |
125 mL | 70 mm |
£64.80
Pack of 4 |
Estimated Shipment: 25-06-2025
Log in to see stock. |
|||||
10700242
![]() |
Thermo Scientific™
2117-0250 |
250 mL | 70 mm |
£77.95
Pack of 4 |
In Stock
Log in to see stock. |
|||||
10149700
![]() |
Thermo Scientific™
2117-0500 |
500 mL | 120 mm |
£129.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
10067820
![]() |
Thermo Scientific™
2117-1000 |
1000 mL | 120 mm |
£154.00
Pack of 4 |
In Stock
Log in to see stock. |
|||||
Description
Protect against breakage and contamination with Thermo Scientific™ Nalgene™; Wide-Mouth Straight-Sided PMP Jars with White Polypropylene Screw Closure. These make excellent museum jars offering good clarity with excellent chemical resistance. They are lighter in weight than glass and lose only 2% of formaldehyde per year. Not liquid leakproof.
Features:- 125mL to 1L jars have white polypropylene closures with recessed tops for stacking.
- Good clarity with excellent chemical resistance make these jars versatile in your lab.
- Straight sides provide full-access and complete sample retrieval.
- 125 and 250mL sizes fit Osterizer™ and other blenders that have Mason Jar threads.
- They are not liquid leakproof, not recommended for liquid transport.
- Chemically compatible with a variety of laboratory chemicals and reagents. Consult the Nalgene Chemical Resistance Guide for specific chemical compatibility information.
- Autoclavable/Transparent
Note: Completely disengage threads or remove cap before autoclaving.
Recycling Information:
Please recycle the corrugate cartons and LDPE bags in which your Nalgene products are packaged.
.
Specifications
Clear | |
PMP | |
Clear | |
Round | |
Screw Cap |
Screw Closure | |
4 | |
Wide | |
Polypropylene |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.