missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nalgene™ Polypropylene Jars with Cover
GREENER_CHOICE

Store and transport solids with these polypropylene jars that are chemically compatible with a wide range of lab materials.

£68.85 - £76.75

Specifications

Color Translucent
For Use With (Application) Storage and Transport
Includes Cover
Material Polypropylene
No. per Case 6
View More Specs

Products 2
Product Code Brand Capacity (Metric) Price Quantity & Availability  
Product Code Brand Capacity (Metric) Price Quantity & Availability  
10656011
GREENER_CHOICE
View Documents
Thermo Scientific™
5352-0002
2.2 L
£68.85
Each
In Stock
to see stock.
Edge
Add to basket
 
10641912
GREENER_CHOICE
View Documents
Thermo Scientific™
5352-0004
4.6 L
£76.75
Each
In Stock
to see stock.
Add to basket
 
Description

Description

Store and transport solids with Thermo Scientific™ Nalgene™ Polypropylene Jars. These jars are chemically compatible with a wide range of lab materials.

  • Straight sides and no neck making access and retrieval easy
  • Larger sizes make excellent waste crocks
  • Autoclavable
Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.
Specifications

Specifications

Translucent
Cover
6
Translucent
Jar
Storage and Transport
Polypropylene
1
Round
Lid
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Thermo Scientific™ Nalgene™ Polypropylene Jars with Cover

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.