Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures
GREENER_CHOICE
Product Code. 10644212
Click to view available options
Product Type:
Replacement Cap
Screw Cap
Spout Cap
Stem and Draw Tube
Closure Size:
20 mm
24 mm
28 mm
38 mm
43 mm
53 mm
63 mm
Color:
Amber
Natural
White
Este artículo no se puede devolver. Vea la política de devoluciones
Este artículo no se puede devolver. Vea la política de devoluciones
Fit these replacement closures onto Thermo Scientific™ Nalgene™ narrow-mouth and wide-mouth bottles.
Fit Thermo Scientific™ Nalgene™ Narrow-Mouth and Wide-Mouth Bottle Replacement Closures onto Nalgene narrow-mouth and wide-mouth bottles.
  • made to fit Nalgene narrow-mouth and wide-mouth bottles
TRUSTED_SUSTAINABILITY
Especificaciones
Color Natural
Material Polypropylene
Closure Included Yes
Mouth Narrow
Shape Round
Product Type Spout Cap
Closure Material Polypropylene
Closure Size 24 mm
Closure Type Spouted Cap
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.