Learn More
Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Economy Bottles

Conduct light-duty, general purpose applications where a heavier bottle is not required with these narrow-mouth amber HDPE bottles.
£87.37 - £141.00
Specifications
Color | Opaque Amber |
---|---|
Description | Amber Narrow-Mouth Economy Bottle |
Material | HDPE |
Sterility | Non-sterile |
Closure Color | Amber |
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Closure Size | Price | Quantity & Availability | ||||||
10020741
|
Thermo Scientific™
DS2085-0001 |
30 mL | 20 mm | N/A | N/A | N/A | |||||
10515021
![]() |
Thermo Scientific™
DS2085-0004 |
125 mL | 24 mm |
£87.37
Pack of 72 |
Estimated Shipment: 17-06-2025
Log in to see stock. |
||||||
10488222
|
Thermo Scientific™
DS2085-0016 |
500 mL | 28 mm | N/A | N/A | N/A | |||||
10459952
![]() |
Thermo Scientific™
DS2085-0032 |
1 L | 38-430 mm |
£141.00
Pack of 24 |
In Stock
Log in to see stock. |
||||||
Description
Conduct light-duty, general purpose applications where a heavier bottle is not required with Thermo Scientific™ Nalgene™ Narrow-Mouth Amber HDPE Bottles. Lightweight versions of the original Nalgene Amber Lab Quality bottle, these high-density polyethylene bottles reduce UV light transmissions to protect light-sensitive liquids.
- Meets U.S. Pharmacopoeia 23 requirements for light transmission
- Excellent chemical resistance to most acids, bases and alcohols make these versatile bottles where exposure to light needs to be restricted
- Economical and recyclable; designed for single use
- Can be used in freezer to -100°C (-148°F) for long-term storage
- Linerless caps provide the ultimate in leakproof† protection without the use of a liner that can wrinkle, cause leaks or contaminate your reagents
- Leakproof† when used with Nalgene closures
- Bottle and carton are readily recyclable in most communities
- Manufactured from non-toxic materials
Note:
Nalgene LDPE, HDPE, PP and PMP containers can be used for long-term storage, but direct UV exposure should be avoided.
.
Recycling Information:
Nalgene reusable HDPE bottles are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Nalgene HDPE bottles and their packaging are readily recyclable. Please recycle the corrugate cartons and LDPE bags in which your Nalgene bottles products are packaged. And at end of product life, please wash and recycle your Nalgene HDPE bottles.
Specifications
Opaque Amber | |
HDPE | |
Amber | |
Yes | |
Round | |
Polypropylene |
Amber Narrow-Mouth Economy Bottle | |
Non-sterile | |
Meets the requirements for light-resistant containers per USP latest edition | |
Narrow | |
Unlined | |
Screw Cap |
† The term leakproof applies to Thermo Scientific Nalgene products that meet the following criteria: a) Bottle/flask/funnel closure systems with closures smaller than 100mm, after they are filled with water, inverted, withstand air pressure of 2psig for 2 minutes, and no water escapes;. b) Bottle/funnel/flask closure systems of larger than 100mm after they are filled with water, inverted for 15 minutes, and no water escapes. Note: these tests, using other liquids, may not yield the same results. To ensure safe usage, customers are advised to test Thermo Scientific Nalgene bottles and closures under conditions of their planned applications. Thermo Scientific Nalgene products are leakproof at ambient temperature and pressure when used with their Nalgene closures.
Your input is important to us. Please complete this form to provide feedback related to the content on this product.