Learn More
N-Phenyldiethanolamine, 97%, Thermo Scientific Chemicals
£15.68 - £77.40
Chemical Identifiers
CAS | 120-07-0 |
---|---|
Molecular Formula | C10H15NO2 |
Molecular Weight (g/mol) | 181.24 |
MDL Number | MFCD00002845 |
InChI Key | OJPDDQSCZGTACX-UHFFFAOYSA-N |
Synonym | n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline |
PubChem CID | 8416 |
IUPAC Name | 2-[N-(2-hydroxyethyl)anilino]ethanol |
SMILES | OCCN(CCO)C1=CC=CC=C1 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11398825
|
Thermo Scientific Alfa Aesar
L13931.22 |
100 g |
£15.68
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11308835
|
Thermo Scientific Alfa Aesar
L13931.36 |
500 g |
£27.80
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11318835
|
Thermo Scientific Alfa Aesar
L13931.0E |
2500 g |
£77.40
2.50kg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
N-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsN-Phenyldiethanolamine is used as phenylephrine intermediates, used as dye intermediates.
Solubility
Soluble in water 3g/L @ 20˚C.
Notes
Air sensitive & hygroscopic. Store under inert gas. Store away from oxidizing agents, air, water/ moisture.
Chemical Identifiers
120-07-0 | |
181.24 | |
OJPDDQSCZGTACX-UHFFFAOYSA-N | |
8416 | |
OCCN(CCO)C1=CC=CC=C1 |
C10H15NO2 | |
MFCD00002845 | |
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline | |
2-[N-(2-hydroxyethyl)anilino]ethanol |
Specifications
120-07-0 | |
1.1 | |
270°C | |
Amine-like | |
C6H5N(CH2CH2OH)2 | |
2096832 | |
n-phenyldiethanolamine, 2,2'-phenylimino diethanol, phenyldiethanolamine, diethanolaniline, diethanolphenylamine, diethanolaminobenzene, phenyl diethanolamine, n,n-diethanolaniline, n,n-dioxyethylaniline, n,n-bis 2-hydroxyethyl aniline | |
OJPDDQSCZGTACX-UHFFFAOYSA-N | |
2-[N-(2-hydroxyethyl)anilino]ethanol | |
8416 | |
97% |
55°C to 59°C | |
8.2 | |
178°C (352°F) | |
C10H15NO2 | |
MFCD00002845 | |
Air sensitive; Hygroscopic | |
Soluble in water 3g/L @ 20?C. | |
OCCN(CCO)C1=CC=CC=C1 | |
181.24 | |
181.23 | |
N-Phenyldiethanolamine |
Safety and Handling
GHS H Statement
H318-H373-H302
Causes serious eye damage.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
P260-P264b-P270-P272-P280-P302+P352-P305+P351+P338-P307+P311-P310-P333+P313-P363-P501c
H317-H318-H370
missing translation for 'einecsNumber' : 204-368-5
missing translation for 'rtecsNumber' : KM2100000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only