missing translation for 'onlineSavingsMsg'
Learn More

n-Octyltriethoxysilane, 95%, Thermo Scientific Chemicals

£27.80 - £146.73

Chemical Identifiers

CAS 2943-75-1
Molecular Formula C14H32O3Si
Molecular Weight (g/mol) 276.49
MDL Number MFCD00039883
InChI Key MSRJTTSHWYDFIU-UHFFFAOYSA-N
Synonym n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
PubChem CID 76262
IUPAC Name triethoxy(octyl)silane
SMILES CCCCCCCC[Si](OCC)(OCC)OCC
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11301577
View Documents
Thermo Scientific Alfa Aesar
L04407.18
50 g
£27.80
50g
Please to purchase this item. Need a web account? Register with us today!
11311577
View Documents
Thermo Scientific Alfa Aesar
L04407.30
250 g
£60.70
250g
Please to purchase this item. Need a web account? Register with us today!
11321577
View Documents
Thermo Scientific Alfa Aesar
L04407.0B
1000 g
£146.73
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

2943-75-1
276.49
MSRJTTSHWYDFIU-UHFFFAOYSA-N
76262
CCCCCCCC[Si](OCC)(OCC)OCC
C14H32O3Si
MFCD00039883
n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
triethoxy(octyl)silane

Specifications

265°C
2943-75-1
MFCD00039883
109°C (228°F)
MSRJTTSHWYDFIU-UHFFFAOYSA-N
triethoxy(octyl)silane
76262
95%
Moisture sensitive
n-Octyltriethoxysilane
<-40°C
C14H32O3Si
2325287
n-octyltriethoxysilane, octyltriethoxysilane, triethoxy octyl silane, silane, triethoxyoctyl, dynasylan octeo, octyl triethoxy silane, prosil 9202, silquest a 137, prosil 9234, a 137 coupling agent
CCCCCCCC[Si](OCC)(OCC)OCC
276.49
276.49
1.417
0.878
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

P264b-P280-P302+P352-P305+P351+P338-P332+P313-P362

H315-H319

missing translation for 'einecsNumber' : 220-941-2

missing translation for 'rtecsNumber' : VV6695500

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.