Learn More
N,N-Dimethyl-4-nitroaniline, 98+%, Thermo Scientific Chemicals
£22.30 - £62.30
Chemical Identifiers
CAS | 100-23-2 |
---|---|
Molecular Formula | C8H10N2O2 |
Molecular Weight (g/mol) | 166.18 |
MDL Number | MFCD00014712 |
InChI Key | QJAIOCKFIORVFU-UHFFFAOYSA-N |
Synonym | 4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro |
PubChem CID | 7491 |
IUPAC Name | N,N-dimethyl-4-nitroaniline |
SMILES | CN(C)C1=CC=C(C=C1)[N+](=O)[O-] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11360024
|
Thermo Scientific Alfa Aesar
L00404.06 |
5 g |
£22.30
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11370024
|
Thermo Scientific Alfa Aesar
L00404.14 |
25 g |
£62.30
25g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
It is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is an aromatic intermediate and in the experimental and theoretical study of the structure of N,N-dimethyl-4-nitroaniline derivatives as model compounds for non-linear optical organic materials.
Solubility
Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency).
Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
100-23-2 | |
166.18 | |
QJAIOCKFIORVFU-UHFFFAOYSA-N | |
7491 | |
CN(C)C1=CC=C(C=C1)[N+](=O)[O-] |
C8H10N2O2 | |
MFCD00014712 | |
4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro | |
N,N-dimethyl-4-nitroaniline |
Specifications
100-23-2 | |
C8H10N2O2 | |
UN2811 | |
4-nitro-n,n-dimethylaniline, benzenamine, n,n-dimethyl-4-nitro, 4-nitrodimethylaniline, p-nitro-n,n-dimethylaniline, p-nitrodimethylaniline, 1-dimethylamino-4-nitrobenzene, 4-dimethylamino nitrobenzene, p-dimethylamino nitrobenzene, n,n-dimethyl-p-nitroaniline, aniline, n,n-dimethyl-p-nitro | |
QJAIOCKFIORVFU-UHFFFAOYSA-N | |
N,N-dimethyl-4-nitroaniline | |
7491 | |
≥98% |
163°C to 166°C | |
MFCD00014712 | |
638087 | |
Soluble in dichloromethane. Insoluble in water. Solubility in hot methanol (almost transparency). | |
CN(C)C1=CC=C(C=C1)[N+](=O)[O-] | |
166.18 | |
166.18 | |
N,N-Dimethyl-4-nitroaniline |
Safety and Handling
GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P305+P351+P338-P311-P312-P330-P332+P313-P361-P363-P501c
H301+H311+H331-H315-H319-H335-H500
missing translation for 'dotInformation' : Hazard Class: 6.1; Packaging Group: II
missing translation for 'einecsNumber' : 202-832-1
missing translation for 'rtecsNumber' : BX7035000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.