missing translation for 'onlineSavingsMsg'
Learn More

N,N-Diethylpropargylamine, 97%, Thermo Scientific Chemicals

Product Code. 15841012
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4079-68-9
Molecular Formula C7H13N
Molecular Weight (g/mol) 111.18
InChI Key JZJXKEWVUBVOEH-UHFFFAOYSA-N
PubChem CID 20010

Specifications

Density 0.8110g/mL
Boiling Point 118°C to 120°C
Flash Point 20°C
Assay Percent Range 96% min. (GC)
Quantity 25 g
Specific Gravity 0.811
Formula Weight 111.18
Percent Purity 97%
Chemical Name or Material N,N-Diethylpropargylamine

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'