missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-N-[(5-methylthien-2-yl)methyl]amine monohydrochloride, 97%, Thermo Scientific™

£47.20 - £128.00

Chemical Identifiers

CAS 912569-78-9
Molecular Formula C7H12ClNS
Molecular Weight (g/mol) 177.69
MDL Number MFCD07106798
InChI Key OQOUPFNJXCXICY-UHFFFAOYSA-N
Synonym n-methyl-1-5-methylthiophen-2-yl methanamine hydrochloride, methyl 5-methylthiophen-2-yl methyl amine hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine hydrochloride, n-methyl-1-5-methyl-2-thienyl methanamine hydrochloride, 2-methyl-5-methylamino methyl thiophene hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine monohydrochloride, aronis23825, methyl 5-methyl 2-thienyl methyl amine, chloride, 2-methyl-5-methylamino methyl thiophene hcl, 2-thiophenemethanamine,n,5-dimethyl-,hydrochloride 1:1
PubChem CID 17290686
IUPAC Name N-methyl-1-(5-methylthiophen-2-yl)methanamine;hydrochloride
SMILES CC1=CC=C(S1)CNC.Cl
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10692325
SDS View Documents
Thermo Scientific Maybridge
CC65096CB
250 mg Amber Glass Bottle
£47.20
250mg
Please to purchase this item. Need a web account? Register with us today!
10327554
SDS View Documents
Thermo Scientific Maybridge
CC65096DA
1 g Amber Glass Bottle
£128.00
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

912569-78-9
177.69
OQOUPFNJXCXICY-UHFFFAOYSA-N
17290686
CC1=CC=C(S1)CNC.Cl
C7H12ClNS
MFCD07106798
n-methyl-1-5-methylthiophen-2-yl methanamine hydrochloride, methyl 5-methylthiophen-2-yl methyl amine hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine hydrochloride, n-methyl-1-5-methyl-2-thienyl methanamine hydrochloride, 2-methyl-5-methylamino methyl thiophene hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine monohydrochloride, aronis23825, methyl 5-methyl 2-thienyl methyl amine, chloride, 2-methyl-5-methylamino methyl thiophene hcl, 2-thiophenemethanamine,n,5-dimethyl-,hydrochloride 1:1
N-methyl-1-(5-methylthiophen-2-yl)methanamine;hydrochloride

Specifications

912569-78-9
100.0
C7H12ClNS
n-methyl-1-5-methylthiophen-2-yl methanamine hydrochloride, methyl 5-methylthiophen-2-yl methyl amine hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine hydrochloride, n-methyl-1-5-methyl-2-thienyl methanamine hydrochloride, 2-methyl-5-methylamino methyl thiophene hydrochloride, n-methyl-n-5-methylthien-2-yl methyl amine monohydrochloride, aronis23825, methyl 5-methyl 2-thienyl methyl amine, chloride, 2-methyl-5-methylamino methyl thiophene hcl, 2-thiophenemethanamine,n,5-dimethyl-,hydrochloride 1:1
CC1=CC=C(S1)CNC.Cl
177.69
177.7
N-Methyl-N-[(5-methylthien-2-yl)methyl]amine monohydrochloride
96.5
97%
MFCD07106798
OQOUPFNJXCXICY-UHFFFAOYSA-N
N-methyl-1-(5-methylthiophen-2-yl)methanamine;hydrochloride
17290686
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if swallowed.
Causes serious eye irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.