missing translation for 'onlineSavingsMsg'
Learn More

N-Methyl-2-(morpholinosulfonyl)benzylamine, 97%, Thermo Scientific™

£40.30 - £98.90

Chemical Identifiers

CAS 941717-08-4
Molecular Formula C12H18N2O3S
Molecular Weight (g/mol) 270.35
MDL Number MFCD09879987
InChI Key ASGSHKVYZRSASF-UHFFFAOYSA-N
Synonym n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
PubChem CID 24229774
IUPAC Name N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine
SMILES CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10163274
SDS View Documents
Thermo Scientific Maybridge
CC69646CB
250 mg Amber Glass Bottle
£40.30
250mg
Please to purchase this item. Need a web account? Register with us today!
10412264
SDS View Documents
Thermo Scientific Maybridge
CC69646DA
1 g Amber Glass Bottle
£98.90
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

941717-08-4
270.35
ASGSHKVYZRSASF-UHFFFAOYSA-N
24229774
CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
C12H18N2O3S
MFCD09879987
n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine

Specifications

941717-08-4
100.0
C12H18N2O3S
n-methyl-2-morpholinosulfonyl benzylamine, 4-2-methylamino methyl phenylsulphonyl morpholine, methyl 2-morpholine-4-sulfonyl phenyl methyl amine, 4-2-methylamino methyl phenylsulfonyl morpholine, n-methyl-1-2-morpholinosulfonyl phenyl methanamine, n-methyl-1-2-morpholine-4-sulfonyl phenyl methanamine
CNCC1=CC=CC=C1S(=O)(=O)N1CCOCC1
270.35
270.35
N-methyl-2-(morpholinosulfonyl)benzylamine
96.5
97%
MFCD09879987
ASGSHKVYZRSASF-UHFFFAOYSA-N
N-methyl-1-(2-morpholin-4-ylsulfonylphenyl)methanamine
24229774
97%
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.