missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals N-Carbobenzyloxy-L-serine, 99+%
£22.40 - £70.40
Chemical Identifiers
CAS | 1145-80-8 |
---|---|
Molecular Formula | C11H13NO5 |
Molecular Weight (g/mol) | 239.23 |
MDL Number | MFCD00002662 |
InChI Key | GNIDSOFZAKMQAO-UHFFFAOYNA-N |
Synonym | z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine |
PubChem CID | 100310 |
IUPAC Name | (2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid |
SMILES | OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1145-80-8 | |
239.23 | |
GNIDSOFZAKMQAO-UHFFFAOYNA-N | |
100310 | |
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O |
C11H13NO5 | |
MFCD00002662 | |
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine | |
(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid |
Specifications
1145-80-8 | |
100.0 | |
White to Cream | |
99% min. (HPLC) | |
HOCH2CH(NHCO2CH2C6H5)CO2H | |
06, IV, 2374 | |
z-ser-oh, n-cbz-l-serine, benzyloxycarbonylserine, z-l-serine, carbobenzyloxy-l-serine, n-carbobenzyloxy-l-serine, cbz-l-ser-oh, n-carbobenzoxy-l-serine, n-benzyloxycarbonyl-l-serine, cbz-l-serine | |
GNIDSOFZAKMQAO-UHFFFAOYNA-N | |
(2S)-3-hydroxy-2-(phenylmethoxycarbonylamino)propanoic acid | |
100310 | |
99+% | |
N-Carbobenzyloxy-L-serine, 99+% |
99.0 | |
115°C to 120°C | |
Authentic | |
C11H13NO5 | |
MFCD00002662 | |
+ 6 | |
Solubility in water: .. Other solubilities: soluble in most common organic solvents | |
OCC(NC(=O)OCC1=CC=CC=C1)C(O)=O | |
239.23 | |
239.23 | |
Powder |
Safety and Handling
missing translation for 'einecsNumber' : 214-546-4
Product Identifier
- N-Carbobenzyloxy-L-serine
RUO – Research Use Only