Learn More
N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline, 97%, Thermo Scientific™
£103.00 - £315.00
Chemical Identifiers
CAS | 1041583-33-8 |
---|---|
Molecular Formula | C14H11F4N |
Molecular Weight (g/mol) | 269.24 |
MDL Number | MFCD12618204 |
InChI Key | JCWUQELJSDRKEP-UHFFFAOYSA-N |
Synonym | n-4-fluorobenzyl-2-trifluoromethyl aniline, n-4-fluorophenyl methyl-2-trifluoromethyl aniline |
PubChem CID | 29292080 |
IUPAC Name | N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline |
SMILES | FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1 |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
NotesStore away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
Chemical Identifiers
1041583-33-8 | |
269.24 | |
JCWUQELJSDRKEP-UHFFFAOYSA-N | |
29292080 | |
FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1 |
C14H11F4N | |
MFCD12618204 | |
n-4-fluorobenzyl-2-trifluoromethyl aniline, n-4-fluorophenyl methyl-2-trifluoromethyl aniline | |
N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline |
Specifications
1041583-33-8 | |
MFCD12618204 | |
JCWUQELJSDRKEP-UHFFFAOYSA-N | |
N-[(4-fluorophenyl)methyl]-2-(trifluoromethyl)aniline | |
29292080 | |
97% |
C14H11F4N | |
n-4-fluorobenzyl-2-trifluoromethyl aniline, n-4-fluorophenyl methyl-2-trifluoromethyl aniline | |
FC1=CC=C(CNC2=CC=CC=C2C(F)(F)F)C=C1 | |
269.24 | |
269.24 | |
N-(4-Fluorobenzyl)-2-(trifluoromethyl)aniline |
Safety and Handling
GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures