missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYO1B Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£135.00 - £349.00
Specifications
Antigen | MYO1B |
---|---|
Concentration | 0.29 mg/mL |
Applications | Immunohistochemistry (Paraffin) |
Classification | Polyclonal |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16803984
|
Proteintech
15012-1-AP-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16893974
|
Proteintech
15012-1-AP-150UL |
150 μL |
£349.00
150µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Motor protein that may participate in process critical to neuronal development and function such as cell migration, neurite outgrowth and vesicular transport.Specifications
MYO1B | |
Immunohistochemistry (Paraffin) | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
O43795, P46735, Q05096 | |
117057, 17912, 4430 | |
MYO1B Fusion Protein Ag6895 | |
Primary | |
-20°C | |
MYO1B |
0.29 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
MMI alpha, MMIa, MYH 1c, MYO1B, Myosin I alpha, myosin IB, myr1 | |
Myo1b | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |