missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MYC Rabbit anti-Human, Clone: 2B17, Proteintech
Rabbit Recombinant Antibody
£135.00 - £360.00
Specifications
Antigen | MYC |
---|---|
Clone | 2B17 |
Concentration | 1000 μg/mL |
Applications | Western Blot, ELISA |
Conjugate | Unconjugated |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17689796
|
Proteintech
80845-1-RR-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
17679796
|
Proteintech
80845-1-RR-100UL |
100 μL |
£360.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MYC antibody apecifically detetects MYC antigen in Human Samples. It is validated for Western Blot, ELISA.Specifications
MYC | |
1000 μg/mL | |
Unconjugated | |
Rabbit | |
PBS with 0.02% sodium azide and 50% glycerol pH 7.3. | |
4609 | |
MYC fusion protein Ag1263 | |
Primary | |
Store at -20°C. Stable for one year after shipment. |
2B17 | |
Western Blot, ELISA | |
Liquid | |
Human | |
BC000141 | |
MYC | |
IgG | |
Protein A purification |