missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ Murashige and Skoog Media
All Murashige and Skoog plant cell culture Media are manufactured from high quality chemicals and reagents.
Brand: MP Biomedicals™ 092623120
42.01 GBP valid until 2025-02-21
Use promo code "24115" to get your promotional price.
View more versions of this product
This item is not returnable.
View return policy
Description
- Each formulation is prepared according to the original specification, except where noted
- Finished lots of media are plant cell tested
Specifications
4°C | |
Murashige and Skoog Basal Medium | |
3.75 to 4.75 |
Powder | |
Solution | |
10 x 1 L |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Warning
Hazard Category
- Serious eye damage/eye irritation Category 2
- Oxidising solid Category 3
Hazard Statement
- H272-May intensify fire; oxidiser.
- H319-Causes serious eye irritation.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P220-Keep/Store away from clothing/combustible materials.
- P264-Wash thoroughly after handling.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P337+P313-If eye irritation persists: Get medical advice/attention.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction