Learn More
Thermo Scientific™ Multiskan™ GO Microplate Spectrophotometer
Perform photometric research applications such as DNA, RNA and protein analysis with the Thermo Scientific™ Multiskan™ GO Microplate Spectrophotometer.
POA
Specifications
Accuracy | 1.0% + 0.003 Abs (0-2.0 Abs); 2% (2.0-2.5 Abs) at 450nm |
---|---|
Bandwidth | <2.5nm |
Depth (Metric) | 43cm |
Incubator Temperature | Ambient +2° to 45°C |
Shaking | Linear |
Beschreibung
Perform photometric research applications such as DNA, RNA and protein analysis with the Thermo Scientific™ Multiskan™ GO Microplate Spectrophotometer. The Multiskan GO features a broad wavelength range (including UV area), path length correction and a fast reading speed. Our intuitive user interface lets you run quick measurements directly from the instrument. Or, use our comprehensive, yet easy-to use Thermo Scientific™ SkanIt™ Software for more demanding assays.
Features:- Freely selectable wavelengths from 200 to 1000nm for the demands of various assays
- Both microplate and cuvette reading for any throughput requirements
- Fast plate measurements and a full sample spectrum in less than 10 seconds
- High quality data guaranteed by extensive self diagnostics
- Unique power save function for reduced energy consumption
- Visual internal software on a large color screen for quick measurements
- Easy and logical assay setup for demanding assays
- A selection of multiple operation languages
Compatible with:
The Multiskan GO is compatible with the Thermo Scientific™ μDrop plate.
Recommended for:
DNA and RNA quantitation and purity; Protein assays; Enzyme assays; Kinetic assays; Immunoassays; Cell proliferation and cytotoxicity assays; Apoptosis assays; Reporter gene assays; GPCR assays
Warranty: 1 year
Spezifikation
1.0% + 0.003 Abs (0-2.0 Abs); 2% (2.0-2.5 Abs) at 450nm | |
43cm | |
Linear | |
10.2 in. | |
Xenon Flash lamp | |
SD<0.003 Abs or CV<0.5% at 450nm (Precision mode); SD<0.003 Abs or CV<1% at 450nm (Fast mode) | |
23.8 lb. | |
11.2 in. |
<2.5nm | |
Ambient +2° to 45°C | |
Monochromator | |
26cm | |
Max: <110w; Power Save: <2.5w | |
200 to 1000nm with 1nm steps | |
10.8kg | |
28.5cm |
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.