missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MP Biomedicals™ SPIN Module and Recovery Tubes
Designed for optimal performance, providing the purest DNA faster than ever, and with no matrix carry-over. SPIN Module and Recovery Tubes are applied in molecular biology for their ability to separate particles from solutions in a few seconds.
Specifications
Content And Storage | Room Temperature |
---|---|
Sample Type | Lysate or other suspension |
For Use With (Application) | SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds. |
Product Type | SPIN Module and Recovery Tubes |
Sample Volume | 500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings |
Description
- Spin Module give faster and cleaner separations than traditional methods such as columns, filters, or simple centrifugation
- When used with the GENECLEAN™ or MERmaid™ protocols, they combine the speed, efficiency, and selectivity of the GLASSMILK™ binding reaction with an even more rapid execution of the separation, washing, and elution steps
Specifications
Room Temperature | |
SPIN Module is applied in molecular biology for its ability to separate particles from solutions in a few seconds. | |
500 to 700 μL liquid per centrifugation, Larger volumes require multiple loadings |
Lysate or other suspension | |
SPIN Module and Recovery Tubes |
Unless specified otherwise, for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title